missing translation for 'onlineSavingsMsg'
Learn More

ZL0420, MedChemExpress

Product Code. p-200097110
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494578

Brand: Medchem Express HY112149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZL0420 is a potent and selective bromodomain-containing protein 4 (BRD4) inhibitor with IC50 values of 27 nM against BRD4 BD1 and 32 nM against BRD4 BD2.

ZL0420 is a potent and selective bromodomain-containing protein 4 (BRD4) inhibitor with IC50 values of 27 nM against BRD4 BD1 and 32 nM against BRD4 BD2.

Specifications

Chemical Name or Material ZL0420
CAS 2230496-80-5
Color Orange
Purity Grade Notes Research
Molecular Formula C16H16N4O2
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (421.84 mM; Need ultrasonic)
SMILES OC1=CC(N)=C(/N=N/C2=CC(CCC(N3)=O)=C3C=C2)C=C1C.[(E)]
Molecular Weight (g/mol) 296.32
Formula Weight 296.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.