missing translation for 'onlineSavingsMsg'
Learn More

ZL0420, MedChemExpress

Product Code. 16494578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494578 10 mg 10mg
16447297 5 mg 5mg
16410887 50 mg 50mg
16403146 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494578 Supplier Medchem Express Supplier No. HY112149
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ZL0420 is a potent and selective bromodomain-containing protein 4 (BRD4) inhibitor with IC50 values of 27 nM against BRD4 BD1 and 32 nM against BRD4 BD2.

ZL0420 is a potent and selective bromodomain-containing protein 4 (BRD4) inhibitor with IC50 values of 27 nM against BRD4 BD1 and 32 nM against BRD4 BD2.

Specifications

Chemical Name or Material ZL0420
CAS 2230496-80-5
Color Orange
Purity Grade Notes Research
Molecular Formula C16H16N4O2
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (421.84 mM; Need ultrasonic)
SMILES OC1=CC(N)=C(/N=N/C2=CC(CCC(N3)=O)=C3C=C2)C=C1C.[(E)]
Molecular Weight (g/mol) 296.32
Formula Weight 296.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.