missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZKSCAN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541597
This item is not returnable.
View return policy
Description
Peptide sequence: EVRAPCSPAR GPERSRQRFR GFRYPEAAGP REALSRLREL CGQWLQPEMH Sequence homology: Cow: 79%; Dog: 90%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 79%.
ZKSCAN4 may be involved in the transcriptional activation of MDM2 and EP300 genes.
Specifications
| ZKSCAN4 | |
| Polyclonal | |
| Unconjugated | |
| ZKSCAN4 | |
| AI841875; Gm11272; OTTMUSG00000000421; P1P373C6; P373c6.1; p373c6.1 (novel C2H2 type zinc finger protein); Zfp307; Zinc finger protein 307; zinc finger protein 427; Zinc finger protein with KRAB and SCAN domains 4; zinc finger with KRAB and SCAN domains 4; ZKSCAN4; ZNF307; ZNF427; ZSCAN36 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 387032 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q969J2 | |
| ZKSCAN4 | |
| Synthetic peptide directed towards the N-terminal of human ZKSCAN4 (aa 38-87). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction