missing translation for 'onlineSavingsMsg'
Learn More

ZKSCAN3 Antibody (2A5), Novus Biologicals™

Product Code. 18422379 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422379 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422379 Supplier Novus Biologicals Supplier No. H00080317M16

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZKSCAN3 Monoclonal antibody specifically detects ZKSCAN3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZKSCAN3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06118
Gene Alias dJ874C20.1, FLJ33906, ZF47, Zfp47, Zfp-47, Zinc finger and SCAN domain-containing protein 13, Zinc finger protein 306KIAA0426, Zinc finger protein 309ZNF306ZFP306, Zinc finger protein 47 homolog, zinc finger protein with KRAB and SCAN domains 3, zinc finger protein zfp47, zinc finger with KRAB and SCAN domains 3, ZNF309, ZSCAN13
Host Species Mouse
Immunogen ZKSCAN3 (AAH06118, 1 a.a. ∽ 538 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MARELSESTALDAQSTEDQMELLVIKVEEEEAGFPSSPDLGSEGSRERFRGFRYPEAAGPREALSRLRELCRQWLQPEMHSKEQILELLVLEQFLTILPGNLQSWVREQHPESGEEVVVLLEYLERQLDEPAPQVSGVDQGQELLCCKMALLTPAPGSQSSQFQLMKALLKHESVGSQPLQDRVLQVPMLAHGGCCREDAVVASRLTPESQGLLKVEDVALTLTPEWTQQDSSQGNLCRDEKQENHGSLVSLGDEKQTKSRDLPPAEELPEKEHGKISCHLREDIAQIPTCAEAGEQEGRLQRKQKNATGGRRHICHECGKSFAQSSGLSKHRRIHTGEKPYECEECGKAFIGSSALVIHQRVHTGEKPYECEECGKAFSHSSDLIKHQRTHTGEKPYECDDCGRTFSQSCSLLEHHRIHTGEKPYQCSMCGKAFRRSSHLLRHQRIHTGDKNVQEPEQGEAWKSRMESQLENVETPMSYKCNECERSFTQNTGLIEHQKIHTGEKPYQCNACGKGFTRISYLVQHQRSHVGKNILSQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80317
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.