missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZKSCAN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552321
This item is not returnable.
View return policy
Description
Immunogen sequence: NLARRNLSRD NRQENYGSAF PQGGENRNEN EESTSKAETS EDSASRGETT GRSQKEFGEK RDQEGKTGER QQKNPEEKTR KEKRDSGPAI GKDKKTITGE RGPREKGKGL GRSFSLSSNF TTPEEVPTGT KSHRCDECGK CFTR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.
The ZKSCAN1 gene encodes a transcriptional regulator of the KRAB. Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. See ZNF91 (MIM 603971) for general information on zinc finger proteins.
Specifications
| ZKSCAN1 | |
| Polyclonal | |
| Unconjugated | |
| ZKSCAN1 | |
| 5930429A01Rik; 9130423L19Rik; 9230118B16Rik; AI646829; AI788588; C87323; KOX18; LOC100720521; PHZ-37; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 139; Zinc finger protein 36; zinc finger protein 36 (KOX 18); zinc finger protein KOX18; Zinc finger protein with KRAB and SCAN domains 1; zinc finger with KRAB and SCAN domains 1; Zkscan1; ZNF139; ZNF36; Zscan21; ZSCAN33 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17029 | |
| ZKSCAN1 | |
| Recombinant protein corresponding to Human ZKSCAN1. Recombinant protein control fragment (Product #RP-89318). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction