missing translation for 'onlineSavingsMsg'
Learn More

ZKSCAN1 Antibody (2C7), Novus Biologicals™

Product Code. 18353149 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353149 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353149 Supplier Novus Biologicals Supplier No. H00007586M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZKSCAN1 Monoclonal antibody specifically detects ZKSCAN1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZKSCAN1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH22378
Gene Alias KOX189130423L19Rik, PHZ-37, Zinc finger protein 139, Zinc finger protein 36, zinc finger protein 36 (KOX 18), Zinc finger protein KOX18, zinc finger protein with KRAB and SCAN domains 1, zinc finger with KRAB and SCAN domains 1, ZNF139, ZNF36MGC138429
Host Species Mouse
Immunogen ZKSCAN1 (AAH22378, 1 a.a. ~ 563 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MMTAESREATGLSPQAAQEKDGIVIAKVEEEDEEDHMWGQDSTLQDTPPPDPEIFRQRFRRFCYQNTFGPREALSRLKELCHQWLRPEINTKEQILELLVLEQFLSILPKELQVWLQEYRPDSGEEAVTLLEDLELDLSGQQVPGQVHGPEMLARGMVPLDPVQESSSFDLHHEATQSHFKHSSRKPRLLQSRALPAAHIPAPPHEGSPRDQAMASALFTADSQAMVKIEDMAVSLILEEWGCQNLARRNLSRDNRQENYGSAFPQGGENRNENEESTSKAETSEDSASRGETTGRSQKEFGEKRDQEGKTGERQQKNPEEKTRKEKRDSGPAIGKDKKTITGERGPREKGKGLGRSFSLSSNFTTPEEVPTGTKSHRCDECGKCFTRSSSLIRHKIIHTGEKPYECSECGKAFSLNSNLVLHQRIHTGEKPHECNECGKAFSHSSNLILHQRIHSGEKPYECNECGKAFSQSSDLTKHQRIHTGEKPYECSECGKAFNRNSYLILHRRIHTREKPYKCTKCGKAFTRSSTLTLHHRIHARERASEYSPASLDAFGAFLKSCV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7586
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.