missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ZK 200775

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Competitive AMPA/kainate antagonist. In rat cortical membranes, displays high affinity for [3H]-AMPA (Ki = 120 nM) and [3H]-CNQX (Ki = 32 nM) binding sites and low affinity for kainate and NMDA channel-associated binding sites (IC50 values range from 2.5 to 11 μM). Inhibits currents induced by AMPA, kainate and NMDA with IC50 values of 21 nM, 27 nM, and > 1 μM respectively. Displays anxiolytic, anticonvulsant and muscle relaxant activity in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 409.25 |
| Purity | >99% |
| Molecular Weight (g/mol) | 409.258 |
| Chemical Name or Material | ZK 200775 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction