missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ZK 200775
GREENER_CHOICE

Product Code. 18033802
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18033802

Brand: Tocris Bioscience™ 2345/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Competitive non-NMDA iGluR antagonist

Competitive AMPA/kainate antagonist. In rat cortical membranes, displays high affinity for [3H]-AMPA (Ki = 120 nM) and [3H]-CNQX (Ki = 32 nM) binding sites and low affinity for kainate and NMDA channel-associated binding sites (IC50 values range from 2.5 to 11 μM). Inhibits currents induced by AMPA, kainate and NMDA with IC50 values of 21 nM, 27 nM, and > 1 μM respectively. Displays anxiolytic, anticonvulsant and muscle relaxant activity in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161605-73-8
Molecular Formula C14H15F3N3O6P

Specifications

Quantity 10 mg
Formula Weight 409.25
Purity >99%
Molecular Weight (g/mol) 409.258
Chemical Name or Material ZK 200775
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.