Learn More
Tocris Bioscience™ ZK 164015
Potent estrogen receptor antagonist
Specifications
Purity >99%
View More Specs
Description
Description
Potent estrogen receptor silent antagonist. Inhibits 17β-estradiol stimulation of luciferase activity (IC50 = 0.025 μM); potently inhibits the growth of estrogen-sensitive human MCF-7 breast cancer cells in vitro (IC50 ∽ 1 nM).
Specifications
Specifications
>99%