missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ZJ 43

Product Code. 18045892
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18045892

Brand: Tocris Bioscience™ 2675/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutamate carboxypeptidase II and III (NAALADase, NAAG peptidase) inhibitor

ZJ 43 is a potent inhibitor of glutamate carboxypeptidase II and III (GCP II and III/NAAG peptidase/NAALADase) (Ki values are 0.8 and 23 nM respectively) that inhibits the hydrolysis of NAAG (IC50 = 2.4 nM) . Does not directly interact with NMDA or metabotropic glutamate receptors. Reduces neuronal degeneration in a rat model of traumatic brain injury (TBI) and reduces locomotor activity in the PCP-model of schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[[[(1 S)-1-Carboxy-3-methylbutyl]amino]carbonyl]-L-glutamic acid
CAS 723331-20-2
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Target Carboxypeptidase Inhibitors
Molecular Formula C12H20N2O7
Formula Weight Observed MW: 304.3
Purity 95%

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.