missing translation for 'onlineSavingsMsg'
Learn More

Zirconyl Chloride Octahydrate, 98%, Reagent grade, Honeywell™

Product Code. p-7120925 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 15622240

Brand: Honeywell Research Chemicals 2243165G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-92-8
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O

Specifications

Color White
Quantity 5 g
Linear Formula ZrOCl2 · 8H2O
UN Number UN3260
Formula Weight 322.25g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Powder or crystals
Density 1.91g/cm3
Chemical Name or Material Zirconyl Chloride Octahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.