missing translation for 'onlineSavingsMsg'
Learn More

Zirconium wire, 1.14mm (0.045in) dia, 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals

Product Code. 11356938
Change view
Click to view available options
Quantity:
10 m
2 m
50 m
Unit Size:
10m
2m
50m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356938 50 m 50m
11332588 2 m 2m
11342588 10 m 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11356938 Supplier Thermo Scientific Alfa Aesar Supplier No. 014627.H2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 50 m
Odor Odorless
Weight ≈6.64g/m
Form Wire
Percent Purity 99.2%
Assay (metals basis excluding Hf), Hf 4.5% max.
Chemical Name or Material Zirconium wire, 1.14mm (0.045 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.