missing translation for 'onlineSavingsMsg'
Learn More

Zirconium wire, 0.25mm (0.01in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11356547
Click to view available options
Quantity:
50 cm
250 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
23995
CHEBI:33342
zirconium
[Zr]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
23995
CHEBI:33342
zirconium
[Zr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 cm
Percent Purity 99.95%
Odor Odorless
Weight ≈0.32g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Zirconium wire, 0.25mm (0.01 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'