missing translation for 'onlineSavingsMsg'
Learn More

Zirconium wire, 0.25mm (0.01 in.) dia., 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals

Catalog Number 11399498
Click to view available options
Quantity:
10 m
2 m
This item is not returnable. View return policy
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
23995
CHEBI:33342
zirconium
[Zr]
This item is not returnable. View return policy
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
23995
CHEBI:33342
zirconium
[Zr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 m
Percent Purity 99.2%
Odor Odorless
Weight ≈0.32g/m
Form Wire
Assay (metals basis excluding Hf), Hf 4.5% max
Chemical Name or Material Zirconium wire, 0.25mm (0.01 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'