missing translation for 'onlineSavingsMsg'
Learn More

Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Product Code. 11378288
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378288 25 g 25g
11388288 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378288 Supplier Thermo Scientific Alfa Aesar Supplier No. 039490.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for glass, optic and ceramic applications, advanced electronics and in light weight structural components in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-70-3
MDL Number MFCD00210648
Synonym zirconium titanium oxide, tetrahydrate titanium zirconium

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 203.11
Percent Purity 99.5%
Odor Odorless
Assay (metals basis excluding Hf), Hf <75ppm
Chemical Name or Material Zirconium titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.