missing translation for 'onlineSavingsMsg'
Learn More

Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Product Code. 11363408
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363408 100 g 100g
11373408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363408 Supplier Thermo Scientific Alfa Aesar Supplier No. 036322.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium silicate is used for manufacturing refractory materials. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. zirconium silicate is as beads for milling and grinding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-90-6
Molecular Formula H4Si2Zr
Molecular Weight (g/mol) 151.43
MDL Number MFCD00151547
InChI Key ZSONHIWWHDAZQO-UHFFFAOYSA-N
Synonym zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane
PubChem CID 6336892
IUPAC Name bis($l^{2}-silanylidene)zirconium
SMILES [SiH2]=[Zr]=[SiH2]

Specifications

Color Gray
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water. It is insoluble in acids,alkali and aqua regia.
Formula Weight 147.39
Percent Purity 99.5%
Odor Odorless
Density 4.88 g/mL
Assay (metals basis excluding Hf), Hf 4%
Chemical Name or Material Zirconium silicide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.