missing translation for 'onlineSavingsMsg'
Learn More

Zirconium silicate, Thermo Scientific Chemicals

Product Code. 11357278
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357278 2 kg 2kg
11490510 500 g 500g
15401177 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11357278 Supplier Thermo Scientific Alfa Aesar Supplier No. 030477.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium silicate is used for manufacturing refractory materials for applications where resistance to corrosion by alkali materials is required. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. It can be also present in some cements. Another use of zirconium silicate is as beads for milling and grinding. Thin films of zirconium silicate and hafnium silicate produced by chemical vapor deposition. Potential alternative gate dielectric to silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-52-7
Molecular Formula O4SiZr
Molecular Weight (g/mol) 183.31
MDL Number MFCD00085353
InChI Key GFQYVLUOOAAOGM-UHFFFAOYSA-N
Synonym zirconium silicate, hyacinth, zircon, zircon zr sio4, zirconite, zircosil 15, standard sf 200, ultrox 500w, a-pax 45m, zirconium iv silicate
PubChem CID 61775
IUPAC Name zirconium(4+);silicate
SMILES [Zr+4].[O-][Si]([O-])([O-])[O-]

Specifications

Melting Point 2550°C
Color White
Physical Form Powder
Quantity 2 kg
Merck Index 14,10181
Solubility Information Insoluble in water,acids,alkali and aqua regia.
Formula Weight 183.3
Odor Odorless
Density 4.56 g/mL
Chemical Name or Material Zirconium silicate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.