missing translation for 'onlineSavingsMsg'
Learn More

Zirconium powder, APS 2-3 micron, packaged in water, Thermo Scientific Chemicals

Catalog Number p-7048254
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
23995
CHEBI:33342
zirconium
[Zr]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-67-7
Zr
91.22
MFCD00011303 MFCD00148878
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
23995
CHEBI:33342
zirconium
[Zr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in water
TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN1358
Quantity 100 g
Odor Odorless
Particle Size APS 2 to 3 Micron
Form Powder
Chemical Name or Material Zirconium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'