missing translation for 'onlineSavingsMsg'
Learn More

Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL

Product Code. 11450420
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450420 500 mL 500mL
11312308 50 mL 50mL
11322308 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11450420 Supplier Thermo Scientific Alfa Aesar Supplier No. 014386.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011303

Specifications

Physical Form Liquid
Concentration ZrCl2O in 5% HCl
UN Number UN1789
Solubility Miscible with water
Quantity 500 mL
Grade Specpure
Chemical Name or Material Zirconium plasma standard solution, Zr 10000μg/mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.