missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide Yttrium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals

Product Code. 11398857
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398857 5 Ea. Pack of 5
11308867 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398857 Supplier Thermo Scientific Alfa Aesar Supplier No. 038509.KU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zirconium Oxide Yttrium Oxide Substrate
Name Note Polished One Side
Physical Form Plate
Quantity 5 Ea.
Molecular Formula ZrO2 + 9.5% Y2O3
MDL Number MFCD01868885
Odor Odorless

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.