missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide, catalyst support, monoclinic phase, Thermo Scientific Chemicals

Product Code. 11340468
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11340468

Brand: Thermo Scientific Alfa Aesar 043815.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Typical properties S.A. 90m2/g, median pore diameter 80/400Å, bimodal total pore volume 0.31cc/g, packing density 1.181g/L

Zirconium oxide is used as a catalyst carrier, diamond simulant, refractory material, pigment, abrasives and solid electrolyte in electrochromic devices. It acts as a precursor to the electroceramic lead zirconate titanate. It is also used in insulation, oxygen sensors, enamels and fuel cell. It finds application to make ceramic knives due to its hardness nature. It is utilized for dental restorations like crowns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011310

Specifications

Physical Form Pellets
Quantity 100 g
Solubility Information Soluble in hot-concentrated hydrogen fluoride and sulfuric acid. Insoluble in water.
Chemical Name or Material Zirconium oxide, catalyst support, 1/8 in. Pellets

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.