missing translation for 'onlineSavingsMsg'
Learn More

Zirconium nitride, 99.5% (metals basis excluding Hf), Hf <3%, Thermo Scientific Chemicals

Product Code. 11326508
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326508 25 g 25g
11336508 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11326508 Supplier Thermo Scientific Alfa Aesar Supplier No. 012138.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium nitride is used in cermets, refractories, and laboratory crucibles. It also used for coating medical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25658-42-8
Molecular Formula NZr
Molecular Weight (g/mol) 105.23
MDL Number MFCD00049642
InChI Key ZVWKZXLXHLZXLS-UHFFFAOYSA-N
Synonym zirconium nitride, zirconium nitride zrn, zirconium nitride,-325 mesh, zirconium nitride trace metals basis excluding hf 25g, zirconium nitride, powder, pore size 1-2 mum trace metals basis
PubChem CID 94359
IUPAC Name azanylidynezirconium
SMILES N#[Zr]

Specifications

Melting Point 2980°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.5% (metals basis excluding Hf)
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Formula Weight 105.23
Density 7.09 g/mL
Chemical Name or Material Zirconium nitride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.