missing translation for 'onlineSavingsMsg'
Learn More

Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals

Product Code. 11460927
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11460927

Brand: Thermo Scientific Alfa Aesar B23317.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-76-7
Molecular Formula C16H40O4Zr+4
Molecular Weight (g/mol) 387.716
MDL Number MFCD00048765
InChI Key ASUYWGMDIAPAKI-UHFFFAOYSA-N
Synonym 1-butanol, zirconium 4+ salt 4:1, 1-butanol, zirconium 4++ salt
PubChem CID 122130442
IUPAC Name butan-1-ol;zirconium(4+)
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Zr+4]

Specifications

Quantity 1000 g
Linear Formula [CH3(CH2)3O]4Zr
UN Number UN1120
Beilstein 4132649
Flash Point 38°C (100°F)
Solubility Information Hydrolyzes in water.
Formula Weight 383.68
Concentration or Composition (by Analyte or Components) 80% w/w in 1-butanol
Odor Alcohol-like
Refractive Index 1.466
Sensitivity Moisture sensitive
Density 1.07
Chemical Name or Material Zirconium n-butoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.