missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) tert-butoxide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11368348
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368348 1 g 1g
11348348 5 g 5g
11358348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368348 Supplier Thermo Scientific Alfa Aesar Supplier No. 039547.03
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

It finds it application as a reagent for the preparation of heterometallic tin(II) or lead(II) zirconium alkoxides. It is used as a catalyst in cyanation process. Zirconium tert-butoxide (ZTB) precursor is used to deposit thin films of Zirconia and other zirconium containing films by atomic layer deposition and chemical vapor deposition methods. Zirconium oxide thin films can also be grown at low temperatures, ranging from 150°C to 300°C, by the presence of moisture along with ZTB or by UV-enhanced atomic layer deposition (UV-ALD) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2081-12-1
Molecular Formula C16H40O4Zr
Molecular Weight (g/mol) 387.72
MDL Number MFCD00075085
InChI Key WRMYASGYMABHCC-UHFFFAOYSA-N
Synonym zirconium iv tert-butoxide, 2-methylpropan-2-ol; zirconium
PubChem CID 25203936
IUPAC Name 2-methylpropan-2-ol;zirconium
SMILES CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Zr]

Specifications

Color Yellow
Density 0.985 g/mL
Boiling Point 90°C (5 mmHg)
Flash Point 85°C (185°F)
Linear Formula Zr[OC(CH3)3]4
Quantity 1 g
Beilstein 3681870
Sensitivity Light and moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 383.68
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Zirconium(IV) tert-butoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.