missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, yttria stabilized, 99% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Catalog Number 11353338
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
1314-23-4
O2Zr
123.22
MFCD00011310 MFCD01868884
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
62395
dioxozirconium
O=[Zr]=O
This item is not returnable. View return policy
1314-23-4
O2Zr
123.22
MFCD00011310 MFCD01868884
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
62395
dioxozirconium
O=[Zr]=O

Stabilized with 1.3% yttrium

Instead of lime for the oxyhydrogen light. As organic intermediate and in pharmaceutical applications. Used as raw material for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ca 5000°C
Melting Point ∽2700°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%, metal basis excluding Hf
Solubility Information Insoluble in water. Soluble in hot and concentrated hydrofluoric acid and sulfuric acid.
Chemical Name or Material Zirconium(IV) Oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'