missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium(IV) oxide, 20% in H2O, colloidal dispersion, Thermo Scientific™
Click to view available options
Quantity:
500 g
2.5 kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White |
Physical Form | Particles in Liquid |
Quantity | 500 g |
Linear Formula | ZrO2 |
Merck Index | 14,10180 |
Solubility Information | Soluble in alcohols,hydrofluoric and hot sulphuric acid. Insoluble in water. |
Formula Weight | 123.22 |
Concentration or Composition (by Analyte or Components) | 20% in H2O |
Odor | Odorless |
Chemical Name or Material | Zirconium(IV) oxide, Colloidal dispersion |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'