missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals

Product Code. 11382598
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382598 2 kg 2kg
11362598 50 g 50g
11372598 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11382598 Supplier Thermo Scientific Alfa Aesar Supplier No. 022989.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium(IV) n-propoxide is used as a precursor in the production of zirconia. It is also used to study the thermophysical properties like thermal conductivity and thermal effusivity of hybrid films and thereby adjusting optic properties of the films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23519-77-9
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
SMILES CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

Physical Form Liquid
Quantity 2 kg
UN Number UN1993
Flash Point 28°C (83°F)
Solubility Information Hydrolyzes in waterImmiscible with water.
IUPAC Name propan-1-olate;zirconium(4+)
Formula Weight 327.56
Concentration or Composition (by Analyte or Components) 70% w/w in n-propanol
Refractive Index 1.451
Sensitivity Moisture sensitive
Density 1.044 g/mL
Chemical Name or Material Zirconium(IV) n-propoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.