missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals

Product Code. 11393337
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393337 25 g 25g
11303347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11393337 Supplier Thermo Scientific Alfa Aesar Supplier No. 017400.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Light, Air and Moisture Sensitive

Used in the van Arkel-de Boer refining process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-26-0
Molecular Formula I4Zr
MDL Number MFCD00049641
Synonym zirconium iodide, unii-h6nzc1810v, zirconium iv ; iodide, acmc-1bzrf, zirconium 4+ tetraiodide

Specifications

Melting Point 431° (sublimation)
Color Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 598.84
Sensitivity Air, light, and moisture sensitive
Density 4.85
Chemical Name or Material Zirconium(IV) iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.