missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals

Product Code. 11394939
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394939 50 g 50g
11384939 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394939 Supplier Thermo Scientific Alfa Aesar Supplier No. 086112.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium(IV) fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production. Together with uranium salt, zirconium fluoride can be a component of fuel-coolant in molten salt reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
Molecular Formula F4Zr
MDL Number MFCD00011307
Synonym Zirconium tetrafluoride

Specifications

Melting Point 600°C (sublimation)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Slightly soluble in water; Soluble in hydrofluoric acid
Formula Weight 167.21
Percent Purity 98%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Chemical Name or Material Zirconium (IV) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.