missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

Product Code. p-7049705
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11363497

Brand: Thermo Scientific Alfa Aesar 018859.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium(IV) ethoxide is used as precursor for the synthesis of olefin-functionalized zirconium-containing monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18267-08-8
Molecular Formula C2H5OZr
Molecular Weight (g/mol) 136.28
MDL Number MFCD00015654,MFCD00015654
InChI Key VCYRFVPNTBNCLW-UHFFFAOYSA-N
Synonym zirconium iv ethoxide
PubChem CID 122130910
SMILES [Zr+4].CC[O-]

Specifications

Melting Point 171°C to 173°C
Boiling Point 235°C (5 mmHg)
Quantity 10 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name zirconium(4+) ethanolate
Formula Weight 271.47
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Zirconium(IV) ethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.