missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

Product Code. 11373497
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373497 50 g 50g
11363497 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11373497 Supplier Thermo Scientific Alfa Aesar Supplier No. 018859.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium(IV) ethoxide is used as precursor for the synthesis of olefin-functionalized zirconium-containing monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18267-08-8
Molecular Formula C2H5OZr
Molecular Weight (g/mol) 136.28
MDL Number MFCD00015654,MFCD00015654
InChI Key VCYRFVPNTBNCLW-UHFFFAOYSA-N
Synonym zirconium iv ethoxide
PubChem CID 122130910
SMILES [Zr+4].CC[O-]

Specifications

Melting Point 171°C to 173°C
Boiling Point 235°C (5 mmHg)
Quantity 50 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name ethanolate;zirconium(4+)
Formula Weight 271.47
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Zirconium(IV) ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.