missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) ethoxide, 99%

Product Code. 10367231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367231 25 g Glass bottle 25g
10663352 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10367231 Supplier Thermo Scientific Acros Supplier No. 335450250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 18267-08-8 | C2H5OZr | 136.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18267-08-8
Molecular Formula C2H5OZr
Molecular Weight (g/mol) 136.28
MDL Number MFCD00015654,MFCD00015654
InChI Key VCYRFVPNTBNCLW-UHFFFAOYSA-N
Synonym zirconium iv ethoxide
PubChem CID 122130910
IUPAC Name ethanolate;zirconium(4+)
SMILES [Zr+4].CC[O-]

Specifications

Melting Point 200°C
Color White
Boiling Point 235°C (7.0 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Linear Formula Zr(OC2H5)4
Quantity 25 g
Formula Weight 271.46
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Zirconium(IV) ethoxide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Zirconium(IV) ethoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.