Learn More
Zirconium(IV) chloride, 98%, cont. 1-2% hafnium(IV) chloride, Thermo Scientific Chemicals
Description
Zirconium(IV) chloride catalyzes the trans-hydrostannylation of acetylenes with Tri-n-butyltin hydride. It is used as a superior catalyst for the fries rearrangement of aryl acetates, giving the least hindered ortho-acetyl phenol in high yields at ambient temperatures. In the presence of ZrCl4, chloromethyl ethers are formed under mild conditions from acid chlorides and trioxane or paraformaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zirconium(IV) chloride catalyzes the trans-hydrostannylation of acetylenes with Tri-n-butyltin hydride. It is used as a superior catalyst for the fries rearrangement of aryl acetates, giving the least hindered ortho-acetyl phenol in high yields at ambient temperatures. In the presence of ZrCl4, chloromethyl ethers are formed under mild conditions from acid chlorides and trioxane or paraformaldehyde.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
Specifications
Melting Point | 331°C (sublimation) |
Quantity | 50 g |
UN Number | UN2503 |
Merck Index | 14,10174 |
Solubility Information | Reacts with water. |
Formula Weight | 233.03 |
Percent Purity | 98% |
Sensitivity | Moisture sensitive |
Density | 2.803 |
Assay | cont. 1 to 2% hafnium(IV) chloride |
Afficher plus |
RUO – Research Use Only