missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals

Product Code. 11363548
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363548 25 g 25g
11373548 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363548 Supplier Thermo Scientific Alfa Aesar Supplier No. 039174.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium(IV) 2-ethylhexanoate is utilized as a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium especially zirconium oxide thin films. It is also used as yttria stabilized zirconia films for solid oxide fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2233-42-3
Molecular Formula C32H60O8Zr
Molecular Weight (g/mol) 664.048
MDL Number MFCD00072684
InChI Key OFYFURKXMHQOGG-UHFFFAOYSA-J
Synonym zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate
PubChem CID 6451377
IUPAC Name 2-ethylhexanoate;zirconium(4+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4]

Specifications

Color Amber
Flash Point >93°C (199°F)
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 664.04
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material Zirconium(IV) 2-ethylhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.