missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals

Product Code. 11367677
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367677 250 g 250g
11377677 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11367677 Supplier Thermo Scientific Alfa Aesar Supplier No. 014065.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium(IV) 2,4-pentanedionate is used as a cross-linking agent for oxygen-containing polymers. It is used as a catalyst in organic synthesis for the nitration of o-nitrotoluene using a dinitrogen pentoxide to get 2,4-dinitrotoluenes and 2,6-dinitrotoluenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17501-44-9
Molecular Formula C20H28O8Zr
Molecular Weight (g/mol) 487.66
MDL Number MFCD00000036
InChI Key DFPGFWYWSAYLCW-UHFFFAOYSA-N
Synonym 4-hydroxypent-3-en-2-one; zirconium
PubChem CID 50912253
IUPAC Name 4-hydroxypent-3-en-2-one;zirconium
SMILES [Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 191°C to 195°C
Color White
Odor Weak
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 487.66
Physical Form Crystals
Chemical Name or Material Zirconium(IV) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.