missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Boiling Point | 4300°C |
| Melting Point | 2700°C |
| CAS | 1314-23-4 |
| Color | White |
| pH | 3 to 4 |
| Physical Form | Powder/Solid |
| Quantity | 250 g |
| Molecular Formula | O2Zr |
| MDL Number | MFCD00011310 MFCD01868884 |
| Synonym | zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler |
| Show More |
Safety and Handling
Product Identifier
- Zirconium(IV) oxide
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?