missing translation for 'onlineSavingsMsg'
Learn More

Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11354939
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds an application as rubber additive and fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15461-27-5
Molecular Formula Cl2OZr
Molecular Weight (g/mol) 178.12
MDL Number MFCD00149898
InChI Key IPCAPQRVQMIMAN-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
PubChem CID 159678
SMILES Cl[Zr](Cl)=O

Specifications

Melting Point 400°C (decomposition)
Physical Form Lump
Quantity 50 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water and alcohol.
IUPAC Name oxozirconium;octahydrate;dihydrochloride
Formula Weight 178.13 (Anhydrous)
Sensitivity Hygroscopic
Density 1.91
Chemical Name or Material Zirconium Dichloride Oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'