missing translation for 'onlineSavingsMsg'
Learn More

Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11489463
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489463 2500 g 2500g
11469463 100 g 100g
11479463 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489463 Supplier Thermo Scientific Alfa Aesar Supplier No. A12342.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-92-8
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O

Specifications

Melting Point 150°C (decomposition)
Quantity 2500 g
UN Number UN3260
Merck Index 14,10184
Solubility Information Soluble in water and alcohol.
Formula Weight 322.25 (178.13 Anhydrous)
Percent Purity 98%
Odor Acidic
Sensitivity Hygroscopic
Density 1.91
Chemical Name or Material Zirconium dichloride oxide octahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Zirconium dichloride oxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: dichlorooxo-octahydrate Zirconium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.