missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium Dichloride Oxide Hydrate, 99.9985% (Metal Basis), Thermo Scientific™, Puratronic™
Click to view available options
Quantity:
10 g
50 g
250 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Assay Percent Range | 99.9985%, metal basis |
Solubility Information | Soluble in cold water,methanol,diethyl ether and alcohol.Slightly soluble in hydrochloric acid. Insoluble in organic solvents. |
IUPAC Name | oxozirconium;hydrate;dihydrochloride |
Formula Weight | 178.13 (Anhydrous) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Zirconium Dichloride Oxide Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'