missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Dichloride Oxide Hydrate, 99.9985% (Metal Basis), Thermo Scientific™, Puratronic™

Product Code. p-4782245
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11386148

Brand: Thermo Scientific Alfa Aesar 011135.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15461-27-5
Molecular Formula Cl2OZr
Molecular Weight (g/mol) 178.12
MDL Number MFCD00149898
InChI Key IPCAPQRVQMIMAN-UHFFFAOYSA-L
Synonym zirconyl chloride hydrate, zirconium dichloride oxide hydrate, zirconium, dichlorooxo-, monohydrate, zirconium oxychloride hydrate, cl2ozr.h2o, zirconium dichloride oxide hydrate, puratronic, zirconyl chloride hydrate trace metals basis
PubChem CID 16211499
SMILES Cl[Zr](Cl)=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9985%, metal basis
Solubility Information Soluble in cold water,methanol,diethyl ether and alcohol.Slightly soluble in hydrochloric acid. Insoluble in organic solvents.
IUPAC Name oxozirconium;hydrate;dihydrochloride
Formula Weight 178.13 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Zirconium Dichloride Oxide Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.