missing translation for 'onlineSavingsMsg'
Learn More

Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals

Product Code. 11378707
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378707 250 g 250g
11388707 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11378707 Supplier Thermo Scientific Alfa Aesar Supplier No. 035808.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium carbide commercially used in tool bits for cutting tools. It is used as an abrasive, in cladding, in cermets and incandescent filaments. It finds application in the coating of uranium dioxide and thorium dioxide particles of nuclear fuel as well as in thermo-photovoltaic radiators and field emitter tips and arrays. It is involved in the usage as refractory coatings in nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-14-3
Molecular Formula CZr
Molecular Weight (g/mol) 103.24
MDL Number MFCD00049640
InChI Key MHMXYYDKIQTOHA-UHFFFAOYSA-N
IUPAC Name methanidylidynezirconiumylium
SMILES [C-]#[Zr+]

Specifications

Melting Point 3540°C
Color Gray
Physical Form Powder
Quantity 250 g
UN Number UN3178
Solubility Information Soluble in concentrated sulfuric acid,hydrofluoric acid and nitric acid. Insoluble in water.
Formula Weight 103.23
Percent Purity 99.5%
Odor Odorless
Density 6.73 g/mL
Assay (metals basis excluding Hf), Hf< 200ppm
Chemical Name or Material Zirconium carbide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.