missing translation for 'onlineSavingsMsg'
Learn More

Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific™

Product Code. 15487365
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487365 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15487365 Supplier Thermo Scientific Alfa Aesar Supplier No. 047030.18
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 3000°C
CAS 12045-64-6
Physical Form Powder
Quantity 50 g
Molecular Formula B2Zr
Linear Formula ZrB2
MDL Number MFCD00064648
UN Number UN3178
Solubility Information It is insoluble aq. soln..
InChI Key VWZIXVXBCBBRGP-UHFFFAOYSA-N
SMILES [B].[B].[Zr]
IUPAC Name diboron zirconium
Molecular Weight (g/mol) 112.84
PubChem CID 9812765
Formula Weight 112.84
Percent Purity 99.5%
Assay (metals basis excluding Hf)
Chemical Name or Material Zirconium boride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.