missing translation for 'onlineSavingsMsg'
Learn More

Merck ZipTip™ Pipet Tips

Product Code. 10156390 Shop All Merck Products
Click to view available options
For Use With (Application):
C18 Media
C4 Media
Micro Bed of C18 Media
Strong Cation Exchange Media
Quantity:
8/Pk.
96/Pk.
960/Pk.
Unit Size:
Pack of 8
Pack of 96
Pack of 960
This item is not returnable. View return policy

Product Code. 10156390

Brand: Merck ZTC18S008

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sample preparation and precise liquid handling in one device

A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.

  • Concentrate and purify 1–100μL biological samples
  • Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques

Order Info

Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Length (English) 1.2 in.
Length (Metric) 3cm
For Use With (Application) C18 Media
Tip Style Specialty, Media-Fixed
Quantity 8/Pk.
For Use With (Equipment) Universal Fit
Product Line ZipTip
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.