missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ZIPtide Substrate

Product Code. 10764511
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10764511 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10764511 Supplier Thermo Scientific™ Supplier No. PV5094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A peptide substrate for the Ca/Calmodulin-dependent protein kinase (CaM) family including CAMK1 and DAPK1.

A peptide substrate for the Ca/Calmodulin-dependent protein kinase (CaM) family including CAMK1 and DAPK1. For use with the Adapta™ Universal Kinase Assay (PV5099).

TRUSTED_SUSTAINABILITY

Specifications

Amino Acid Sequence KKLNRTLSFAEPGW
Assay Entry Biochemical ADP accumulation
Content And Storage 2 mM ZIPtide Peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA. Store (at -20°C.)
Detection Method Fluorescence
Quantity 1 mL
Substrate peptide for CaM kinases
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 1612, 8536
Molecular Weight (g/mol) 1646.9 Da
Shipping Condition Dry Ice
Target Entry DAPK1, CAMK1
Validated Kinases CAMK1, DAPK1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.