missing translation for 'onlineSavingsMsg'
Learn More

Ziprasidone, MedChemExpress

Product Code. 16446088
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16446088

Brand: Medchem Express HY14542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ziprasidone (CP-88059), an orally active antipsychotic agent, is a combined 5-HT and dopamine receptor antagonist. Ziprasidone mesylate trihydrate has affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM) and 5-HT1A (Ki=3.4 nM).

Ziprasidone (CP-88059), an orally active antipsychotic agent, is a combined 5-HT and dopamine receptor antagonist. Ziprasidone mesylate trihydrate has affinities for Rat D2 (Ki=4.8 nM), 5-HT2A (Ki=0.42 nM) and 5-HT1A (Ki=3.4 nM).

Specifications

Chemical Name or Material Ziprasidone
CAS 146939-27-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H21ClN4OS
Quantity 10 mg
Synonym CP-88059
Solubility Information DMSO : 13.5 mg/mL (32.69 mM; Need ultrasonic)
SMILES ClC1=CC2=C(CC(N2)=O)C=C1CCN(CC3)CCN3C4=NSC5=C4C=CC=C5
Molecular Weight (g/mol) 412.94
Formula Weight 412.94
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.