missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ziprasidone Hydrochloride Monohydrate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Ziprasidone Hydrochloride Monohydrate
Specifications
Specifications
| Chemical Name or Material | Ziprasidone hydrochloride monohydrate |
| CAS | 138982-67-9 |
| Analyte or Component Names | Ziprasidone hydrochloride monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H21 Cl N4 O S . Cl H . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ziprasidone Hydrochloride Monohydrate |
| InChI Formula | InChI=1S/C21H21ClN4OS.ClH.H2O/c22-17-13-18-15(12-20(27)23-18)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)28-24-21;;/h1-4,11,13H,5-10,12H2,(H,23,27);1H;1H2 |
| SMILES | O.Cl.Clc1cc2NC(=O)Cc2cc1CCN3CCN(CC3)c4nsc5ccccc45 |
| IUPAC Name | 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction