missing translation for 'onlineSavingsMsg'
Learn More

Ziprasidone Hydrochloride Monohydrate 1.0 mg/ml in Methanol (as anhydrous free base), LoGiCal

Product Code. 30463699
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30463699

Brand: LoGiCal LGCAMP1273.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ziprasidone hydrochloride monohydrate
CAS 138982-67-9
Analyte or Component Names Ziprasidone hydrochloride monohydrate
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H21 Cl N4 O S . Cl H . H2 O
Certifications/Compliance ISO 17034
Synonym Ziprasidone Hydrochloride Monohydrate
InChI Formula InChI=1S/C21H21ClN4OS.ClH.H2O/c22-17-13-18-15(12-20(27)23-18)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)28-24-21;;/h1-4,11,13H,5-10,12H2,(H,23,27);1H;1H2
Solution Type Single Solution
SMILES O.Cl.Clc1cc2NC(=O)Cc2cc1CCN3CCN(CC3)c4nsc5ccccc45
IUPAC Name 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 467.41
Quantity 1 mL
Formula Weight 466.0997 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.