missing translation for 'onlineSavingsMsg'
Learn More

Ziprasidone Amino Acid Sodium Salt(Ziprasidone Impurity C), TRC

Product Code. 30449990
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449990 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449990 Supplier Toronto Research Chemicals Supplier No. TRCZ48499525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ziprasidone Amino Acid Sodium Salt(Ziprasidone Impurity C)
CAS 1798033-44-9
Molecular Formula C21H22ClN4NaO2S
Quantity 25 mg
Synonym [2-Amino-5-[2-[4-(1,2-benzisothiazol-3-yl)piperazin-1-yl]ethyl]-4-chlorophenyl]acetic Acid Sodium Salt, 2-Amino-5-[2-[4-(1,2-benzisothiazol-3-yl)-1-
InChI Formula InChI=1S/C21H23ClN4O2S.Na/c22-17-13-18(23)15(12-20(27)28)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)29-24-21;/h1-4,11,13H,5-10,12,23H2,(H,27,28);/q;+1/p-1
SMILES ClC1=CC(N)=C(CC(O[Na])=O)C=C1CCN2CCN(C3=NSC4=C3C=CC=C4)CC2
IUPAC Name sodium 2-(2-amino-5-(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-4-chlorophenyl)acetate
Molecular Weight (g/mol) 452.93
Formula Weight 452.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.