missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX ZIPK Protein, Active

Product Code. 17320424
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17320424

Brand: Eurofins DiscoverX 14608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal, GST tagged, recombinant, of human ZIPK amino acids 1-290, for use in Kinase Assays.

Death-associated protein kinase (DAPK) is a calcium/calmodulin-dependent kinase that phosphorylates serine/threonine residues on proteins. DAPK modulates cell death induced by IFN-g, TNF-a, Fas, and detachment from the extracellular matrix. DAPK has been also implicated in TGF-binduced apoptosis in several cultured cell lines in which Smad proteins mediate transcriptional activation of DAPK. DAPK3, also known as Zipper interacting protein (ZIP) kinase, contains an N-terminal kinase domain and a C-terminal leucine zipperstructure and binds to ATF4 that is a member of ATF/CREB family. Overexpression of ZIP kinase induces apoptosis. ZIP has also been shown to have a potential role in regulating VSMC contraction.

GenBank AB007144

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 64.3 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in E. coli cells
Protein Form Truncated
Purity or Quality Grade ≥51%
Protein ZIPK, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.