Learn More
ZIP4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20625-1-AP-20UL
 View more versions of this product
Product Code. 16870185
  • £135.00 / 20µL
Description
The zinc transporter ZIP4, also known as SLC39A4, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP4 was initially identified as zinc-uptake transporter in the intestine and as a candidate gene for acrodermatitis enteropathica (AE), a rare inherited defect in the absorption of dietary zinc. Similar to other ZIP family members, aberrant expression of ZIP4 has been linked to cancer, specifically the progression and aggressiveness of pancreatic cancer. ZIP4 expression has also been shown to be activated in hepatocellular carcinomas, acting to repress apoptosis, enhance cell cycle and increase migration, suggesting ZIP4 may play a role in multiple types of cancers.
Specifications
Specifications
ZIP4
Polyclonal
Unconjugated
SLC39A4
AEZ, SLC39A4, Zinc transporter ZIP4, ZIP 4, ZIP4, Zrt and Irt like protein 4
Rabbit
Antigen Affinity Chromatography
RUO
55630, 72027
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P5W5, Q78IQ7
SLC39A4
ZIP4 Fusion Protein Ag14524
20 μL
Primary
Mouse, Human